missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ Radiant Heat Cover
Description
Features:
- Modular range allows for multiple combinations and different coverage levels
- Compatible with 3M™ Versaflo™ Helmet
- Modular
- Protects against radiant heat
Certifications
NIOSH Approved
Specifications
Specifications
| Certifications/Compliance | NIOSH Approved |
| Color | Silver |
| Disposable | Reusable |
| Material | Aluminized |
| For Use With (Application) | Foundries |
| For Use With (Equipment) | Versaflo™ M-307 Hard Hat Assembly |
| Model | M-973 N |