missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ PAPR System Filters and Cartridges
Description
- Filters and cartridges work with belt-mounted Versaflo PAPR units
- Recommended for use in healthcare, pharmaceutical, heavy industrial, manufacturing and/or general manufacturing environments
- NIOSH approved
- 3M Catalog# TR-6710N-40
Compliance
NIOSH approved.
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Color | Magenta |
| Filter Type | Particulate |
| Product Type | HE Filter |
| For Use With (Application) | Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing |
| For Use With (Equipment) | TR-600/800 Series |
| Model | TR-6710N-40 |
| Quantity | 10/Pk. |