missing translation for 'onlineSavingsMsg'
Learn More

3M™ TR-300 Versaflo™ PAPR Assemblies with Belt and Battery

Catalog No. 17000723
Change view
Click to view available options
Battery Life:
10 to 12 hours
6 hours
Belt Material:
Easy Clean belt
High durability belt
Battery Type:
High Capacity Lithium Ion
Lithium-ion
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Life Belt Material Battery Type
17000723 10 to 12 hours High durability belt High Capacity Lithium Ion
17000720 10 to 12 hours Easy Clean belt High Capacity Lithium Ion
17000721 6 hours Easy Clean belt Lithium-ion
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17000723 Supplier 3M™ Supplier No. 7100153813

missing translation for 'validMainframeMsg'

Includes

3M™ Versaflo™ PAPR Assembly, High Durability Belt, High Capacity Battery, Battery Charger Kit (TR-341N)

TR-300 series PAPR assemblies with belt and battery provide performance, protection and functionality with a slim, lightweight balanced design. Headtops and breathing tubes sold separately.

Features:

  • Two selectable flow rate options offers more options and comfort to the user
  • Choose from Easy Clean or Industrial assemblies
  • Easy-to-use and versatile PAPR system for particulate environments
  • High efficiency particulate prefilter included (reorder Fisher catalog #19-039-569)
  • Optional nuisance organic vapor filter available
  • 3M Catalog# TR-304N+, TR-305N+, TR-306N+, TR-307N+

Order Info

No Headgear Included.

Specifications

Battery Life 10 to 12 hours
Battery Type High Capacity Lithium Ion
Belt Material High durability belt
Cartridge Type TR-3712N
Certifications/Compliance NIOSH Approved
Color Beige
Filter Type HEPA
Product Type PAPR Assembly, No Headtop
Flow Rate 6.5 CFM and 7.2 CFM
For Use With (Application) Powder Handling
Includes PAPR Unit, High-Durability Belt (TR-326), Battery, Battery Charger Kit (TR-341N), HE Filter, PAPR Filter Cover, Power Supply, Prefilter, Air Flow Indicator, Spark Arrestor
Model TR-306N+
Mounting Type Belt-mounted
Show More Show Less