Learn More
3M™ Versaflo™ M-400 Series PAPR Outer Shroud
Catalog No. 19039529
Change view
Click to view available options
Style:
High Durability
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
19039529 High Durability
19039526 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19039529 Supplier 3M™ Supplier No. 7100289940
Outer shrouds are replacement parts for use with M-400 helmets.
  • The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
  • 3M Catalog# M-445, M-446, M-447, M-448
Specifications
Assigned Protection Factor (APF) 1,000
Material Cordura Nylon
Product Type Outer Shroud
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
For Use With (Equipment) M-400 helmet
Model M-448
Style High Durability