missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ M-300 Series Respiratory Hardhat Assembly

Catalog No. 19039515
Change view
Click to view available options
Model:
M-305
M-307
Includes:
Flame-Resistant Faceseal
Standard Visor and Faceseal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
19039515 M-305 Standard Visor and Faceseal
19039516 M-307 Flame-Resistant Faceseal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19039515 Supplier 3M™ Supplier No. 7000002382

Versaflo™ M-300 Series Respiratory Hardhat Assembly provides respiratory and limited head and eye protection.

Features:

  • Chemical and Scratch Resistant Lenses
  • Large Viewing Area
  • Lightweight, Easy Visor Replacement System

Environmental Conditions:

  • Cold Working Environment
  • High Mobility Required
  • Safety Helmet Required

Hazard Type:

  • Gas, Vapour and Particulates
  • Liquid Splash
  • Low Energy Flying Particles/Dust
  • Falling Objects
  • Sparks and Hot Particles

Applications:

  • Automotive, food and beverage manufacturing
  • Metal production and fabrication
  • Military maintenance
  • Repair and operation
  • Pharmaceutical
  • Transportation

Specifications

Assigned Protection Factor (APF) 25
Certifications/Compliance CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001, EN 397
Color Black
Product Type Respirator Hard Hat
For Use With (Application) Demolition, Food and Beverage Manufacturing, Metalworking, Pulp and Paper, Agriculture, Smelting, Woodworking, Chemical Processing, Construction
For Use With (Equipment) 3M Air Delivery Units
Includes Standard Visor and Faceseal
Model M-305
Sterility Non-sterile
Suspension Type Textile
Visor Type Standard Visor and Faceseal
With Faceseal Yes
Dimensions (L x W x H) 346 x 346 x 254 mm
Length (English) 13.6 in.
Length (Metric) 346 mm
Width (English) 13.6 in.
Width (Metric) 346 mm
Height (English) 9.6 in.
Height (Metric) 254 mm
Weight (English) 1.8 lb.
Weight (Metric) 832 g
Show More Show Less