missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ S-Series Headcovers, Non-Adjustable Fabric Suspension

Catalog No. 19039547
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19039547 Small/Medium
19039546 Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19039547 Supplier 3M™ Supplier No. 7000127462

For use with select series 3M™ powered air purifying and supplied air respirator systems.

Applications:

Automotive, food and beverage manufacturing, Metal production and fabrication, Military maintenance, Repair and operation; Pharmaceutical, Transportation

  • 3M Catalog# S-103L-20, S-103S-20

Specifications

Certifications/Compliance NIOSH
Color White
Material Polypropylene
Product Type Economy Headcover
Size Small/Medium
For Use With (Application) Military maintenance, repair and operation, pharmaceutical, transportation, metal production and fabrication, automotive, food and beverage manufacturing
For Use With (Equipment) GVP, Breathe Easy™ V-Series, TR-300, TR-600, TR-800
Model S-103S-20
Suspension Type Integrated, Non-adjustable Fabric
Visor Type PET