Learn More
3M™ Versaflo™ BT-Series Breathing Tube Accessory, Adapter
Catalog No. 19805609
Change view
Click to view available options
For Use With (Equipment):
Versaflo BT-Series Breathing Tube
Catalog No. For Use With (Equipment)
19805609 Versaflo BT-Series Breathing Tube
1 options
Catalog No. 19805609 Supplier 3M™ Supplier No. 7000002156
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to adapt the V-100, V-200, V-300 and V-400 air regulating valves.
  • Designed to adapt V-100, V-200, V-300, V-400 air regulating valves to accommodate BT-200 and BT-222 breathing tubes
  • 3M Catalog# V-199
Specifications
For Use With (Equipment) Versaflo BT-Series Breathing Tube
Model V-199
Product Type Adapter