Learn More
3M™ Versaflo™ TR-830N Battery
Catalog No. 17000730
Catalog No. 17000730 Supplier 3M™ Supplier No. 7100150925
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
TR-838 Battery Attachment Tool
Versaflo™ lithium-ion battery pack for TR-800 series PAPRs charges quickly, features a charge level indicator and mechanically attaches (with tool) to the TR-802N PAPR blower motor.
Features:
  • Mechanical attachment ensures secure connection as required by the most current UL standard (UL 60079, 6th Edition, 2013) for Intrinsically Safe PAPRs
  • Approximately 750 full charge/discharge cycles
  • Built in charge level indicator
  • Charges quickly (90% charge in approximately 3.5 hours)
  • 3M Catalog# TR-830
Certifications
NIOSH Approved
Compliance
UL-certified Intrinsically Safe (UL 60079, 6th Edition, 2013).
Specifications
Battery Type Lithium-ion
Certifications/Compliance UL-Certified Intrinsically Safe (UL 60079, 6th Edition, 2013)
Product Type Battery Pack
For Use With (Application) Spray Painting, Mixing, Painting, Powder Handling, General Maintenance, Cleaning, Powder Coating, Liquid Chemical Handling
For Use With (Equipment) Versaflo™ TR-800 Series PAPR Systems
Includes TR-838 Battery Attachment Tool
Model TR-830