missing translation for 'onlineSavingsMsg'
Learn More

3M™ Supplied-Air Respirator Compatible Breathing Tubes (Loose Fitting Headgear)

Catalog No. 19021593
Change view
Click to view available options
For Use With (Equipment):
Breathing Tube for H-Series hoods
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19021593 Breathing Tube for H-Series hoods
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19021593 Supplier 3M™ Supplier No. 7000005372

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for use with 3M™ Supplied Air Hood Systems.

For use with the 3M hoods, helmets, headgear and facepieces described in ordering table.

  • Breathing tubes, compressed air supply hoses, and air control devices are required parts of a NIOSH-approved system.
  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

Specifications

Color Gray
Material Vinyl
Product Type Breathing Tube
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
For Use With (Equipment) Breathing Tube for H-Series hoods
Model W-5114