Learn More
3M™ 6000 / 7000 Series Half- and Full Facepiece Accessory, Supplied-Air Dual-Airline Adapter Kit
Catalog No. 189991053
Change view
Click to view available options
:
Back-Mounted; High Pressure
Catalog No.
189991053 Back-Mounted; High Pressure
1 options
Catalog No. 189991053 Supplier 3M™ Supplier No. 7000126161
Only null left
Back mounted
  • Include breathing tube and connector assembly to easily convert full- and half-facepiece respirators for dual airline use
  • 3M Catalog# SA-2000
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Product Type Airline Adapter
For Use With (Equipment) 6000, 7000 Series Full/Half Facepiece Respirator
Includes 3M™ breathing tube sa-2500, 3M™ waist belt gvp-127 and 3M™ valve sa-1009
Model SA-2000