missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Speedglas' Helmet Protection Plates
Description
- Auto-Darkening Filter Protection
- Replacement Part
- 3M Catalog# 04-0260-00, 04-0260-00, 04-0270-04, 04-0280-00, 04-0290-00, 04-0290-00
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | FlexView Welding Helmet System |
| For Use With (Application) | Replacing Helmet Protection Plates |
| Material | Polycarbonate |
| Model | 04-0260-00 |
| Product Type | Protection Plate |