Learn More
3M™ Speedglas™ 9100 MP Welding Helmet Replacement Parts
Compatible with all 9100 (V, X, XX) filters
Supplier: 3M™ 7000128225
Description
For use with 3M Speedglas 9100 MP Welding Helmets
- Assembly
- Blasting
- Cleaning
- Demolition
- Electrical
- Facility Maintenance
- Grinding
- Health and Safety Testing
- Machine Operations
- Painting
- Remediation
- Sanding
- Welding
- 3M Catalog# 27-0099-28, 27-0099-52, 27-0099-53, 27-0099-63, 27-0099-68, 27-0099-72
Compliance
ANSI Z89.1 - 2009, ANSI Z87.1-2010
Specifications
| Inner shield | |
| 27-0099-63 | |
| 1 Ea. | |
| Welding Helmet Replacement Part |