missing translation for 'onlineSavingsMsg'
Learn More

3M™ Speedglas™ 9100 MP Welding Helmet Assemblies

Catalog No. 19164124
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19164124 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19164124 Supplier 3M™ Supplier No. 7000128221

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Face seal
  • Air duct assembly
  • Headband
  • Hard hat
  • Chrome front panel

Compatible with all 9100 (V, X, XX) filters

  • Flip-up assembly for grinding and prep work
  • SideWindows shade 5 side filters that increase peripheral vision
  • Ergonomic, adjustable head suspension for increased comfort, helmet stability and weight distribution
  • Earmuff protection available as optional accessory
  • Hard hat shell is made of heat resistant polycarbonate
  • A wide (∽8 x 4.25 in.), curved, protective visor with excellent views up, down and peripherally
  • Designed from inception to take one of our optional Peltor™ Earmuffs: low-profile hearing protection with liquid/foam ear cushions and orange housings
  • Dark state: shade 5, 8, 9-13 (variable)
  • Light state: shade 3
    • UV/IR protection: shade 13 (permanent)
    • Switching time, light-dark: <0.1ms (+23°C)
      • Assembly
      • Blasting
      • Cleaning
      • Demolition
      • Electrical
      • Facility Maintenance
      • Grinding
      • Health and Safety Testing
      • Machine Operations
      • Painting
      • Remediation
      • Sanding
      • Welding
      • 3M Catalog# 27-0099-35SW

Compliance

ANSI Z89.1 - 2009, ANSI Z87.1-2010

Specifications

Color Black
Includes Includes face seal, air duct assembly, headband, hard hat, chrome front panel
Model 27-0099-35SW
Quantity 1 Ea.
Style Welding helmet