Learn More
3M™ SecureFit™ Safety Glasses with Anti-fog Lens
Catalog No. 17000710
Change view
Click to view available options
Model:
SF401SGAF-BLU
SF401SGAF-BLU-F
SF407SGAF-BLU
Lens Tint:
Clear
Gray
Frame Color:
Blue, Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Lens Tint Frame Color
17000710 SF401SGAF-BLU Clear Blue, Gray
17000711 SF407SGAF-BLU Gray Blue, Gray
17000712 SF401SGAF-BLU-F Clear Blue, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17000710 Supplier 3M™ Supplier No. 7100150838
Features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
Features:
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Scotchgard™ Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated spectacle
  • Polycarbonate lens absorbs 99.9% UVA & UVB rays
  • Scotchgard™ Anti-fog coating provides fog and abrasion resistance.
  • 3M Catalog# SF401SGAF-BLU, SF401SGAF-BLU-F, SF401SGAF-RED, SF402SGAF-BLU, SF402SGAF-BLU-F, SF403SGAF-YEL, SF405SGAF-BLA, SF406SGAF-BLA, SF407SGAF-BLU
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Electrical
Frame Color Blue, Gray
Model SF401SGAF-BLU
No. per Pack 20/Pk.
Temple Style SecureFit™
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Frameless
Show More Show Less