missing translation for 'onlineSavingsMsg'
Learn More

3M™ Rugged Comfort 6500 Series Quick Latch Half Facepiece Reusable Respirator

Catalog No. 19096915
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19096915 Medium
19096914 Small
19096916 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19096915 Supplier 3M™ Supplier No. 7000128239
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rugged Comfort reusable respirator combines comfort, durability, and stability with an added quick latch mechanism to easily enable the mask to drop down from face without removing head straps.

  • Resilient silicone faceseal offers extended facepiece life even in high-heat environments
  • Quick latch design eliminates need to remove a hardhat or faceshield when lowering or raising the mask
  • Sleek, low-profile design
  • Special valve cover helps to direct exhaled breath and moisture downward
  • 3M’s proprietary Cool Flow™ Valve helps to make breathing easier by reducing heat and moisture
  • Fits securely and comfortably on the head
  • Compatible with all 3M™ bayonet-style filters and cartridges:
    • 6000 Series Cartridges
    • 2000, 2200, 7000 or 5000 Series Filters

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Certifications (OSHA) OSHA 29 CFR Part 1920.134
Connection Type Bayonet
Color Gray, Teal
Product Type Half-Mask Respirator
Size Medium
Facepiece Material Silicone
For Use With (Application) Facility maintenance, grinding, machine operations, sanding, welding, demolition, assembly, cleaning
For Use With (Equipment) Line Compressed Air System SA Series, 3M™ 6000 Series cartridges and 2000, 2200, 7000 or 5000 Series filters
Includes Quick Latch
Model 6502QL
Suspension Type Yolk and cradle
Weight (English) 0.37 lb.
Weight (Metric) 0.16 kg
Strap Attachment Type Quick Latch Mechanism
Show More Show Less