missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Breathe-Easy™ Respirator Accessory, Belt Assembly
Description
- Web material with a plastic buckle
- Holds the turbo unit in place
- 3M Catalog# 520-02-90R01
Specifications
Specifications
| Color | Black |
| Disposable | Reusable |
| Material | Nylon |
| Product Type | Belt |
| For Use With (Application) | Assembly, Machine Operation, Used to Hold The Turbo Unit in Place |
| For Use With (Equipment) | Breathe Easy™ PAPR Systems |
| Includes | Plastic buckle |
| Model | 520-02-90R01 |