Learn More
3M™ Push-to-Talk (PTT) Big Button Connector for PELTOR™ ORA TAC In-Ear Communications Headset
Catalog No. 19164109
Change view
Click to view available options
For Use With (Equipment):
PELTOR™ ORA TAC In-Ear Communications Headset
Catalog No. For Use With (Equipment)
19164109 PELTOR™ ORA TAC In-Ear Communications Headset
1 options
Catalog No. 19164109 Supplier 3M™ Supplier No. 7000128208
Large Push-to-Talk button is useful when gloved hands or garments prevent activation of the PTT on the ORA TAC handset.
  • Use in conjunction with the 3M™ PELTOR™ ORA TAC in-ear headset
  • Useful when gloved hands or garments prevent activation of the PTT on the ORA TAC handset
  • Move and clip the Big PTT Button to a convenient location on your body
  • 4.5 in. curly cable (15 in. total unextended length)
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
For Use With (Equipment) PELTOR™ ORA TAC In-Ear Communications Headset
Model CON-SDBB
Product Type Remote Big Button Push-to-Talk (PTT)