Learn More
3M™ Privo™ Safety Eyewear
Offers European styling
Supplier:  3M™ 7000148518
 View more versions of this product
Catalog No. 191-66-815
Only null left
Explore available promotions
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • Lens and temple tip color options
  • Soft-tipped temples
  • Soft comfort nose bridge
  • Durable polycarbonate lenses absorb 99.9% UV
  • 3M Catalog# 12261-00000-20, 12263-00000-20, 12264-00000-20, 12265-00000-20, 12266-00000-20, 12268-00000-20
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
Specifications
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Silver
Red
Gray
12266-00000-20
Polycarbonate
Product Suggestions
Product Suggestions