Learn More
3M™ Polycarbonate Faceshields
Description
Includes
Faceshield only. (Head gear is sold separately.)
- 3M Total Performance faceshields feature dual-coat technology that provides a durable, abrasion-resistant hard coat on the outside and an anti-fog coating on the inside. Made of durable, splash and impact-resistant polycarbonate.
- 3M provides reliable head and face protection products that meet regulated performance and safety requirements.
- 3M Catalog# 82600-00000, 82601-00000, 82602-00000, 82603-00000, 82604-00000, 82705-10000, 82706-10000
- For use with 3M H-Series Hard Hats/Headgear (H24M/7000002292, H24T/7100113761, H18/7000002291, H18-S/7000052752, H8A/7000002290, H10/7000052751, H8A-2/7000127237).
Compliance
Meets high-impact requirements of ANSI Z87.1-2010
Specifications
Specifications
| Thickness (English) | 0.75 in. |
| Lens Tint | Gold-plated, Clear, Gold-plated |
| For Use With (Equipment) | 3M H-Series Hard Hats/Headgear |
| Model | 82602-00000 |
| Lens Material | Polycarbonate |