missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ Optime™ 101 Ear Muffs

Catalog No. p-90969
Change view
Click to view available options
Model:
H7A
H7B
H7P3E
Style:
Behind-the-head
Hard Hat
Over-the-head
Noise Reduction Rating (NRR):
24 dB
26 dB
27 dB
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR)
173881A H7A Over-the-head 27 dB
2705563 H7B Behind-the-head 26 dB
173881B H7P3E Hard Hat 24 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 173881A Supplier 3M™ Supplier No. 7000009669
Only null left

PELTOR™ Optime™ 101 Ear Muffs are recommended for TWA noise exposures up to 101 dBA.

  • For noise exposures up to 101 dBA
  • Liquid-foam ear cushion
  • Ear cushions snap in and out of place for easy cleaning
  • Over-the-Head Style or Hard Hat-Attached styles
  • Adjusts to fit any head size
  • Soft, padded headband for extra comfort
  • Stainless-steel band wires
  • Four-point suspension system distributes suspension evenly

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Color Black, Green
ANSI S3.1974 Yes
Cushion Material Liquid-foam
CSA Z94.2 Class A
Cushion Color Black
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
Model H7A
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 27 dB
Product Type PELTOR™ Optime™
Headband Color Black
Dielectric No
Material Foam, Plastic, Metal
Style Over-the-head
Show More Show Less