missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ LiteCom FRS Headset

Catalog No. 19072314
Change view
Click to view available options
Model:
MT53H7A4602-NA
MT53H7B4602-NA
MT53H7P3E4602-NA
Style:
Hard Hat
Neckband
Over-the-head
Noise Reduction Rating (NRR):
24 dB
25 dB
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR)
19072314 MT53H7A4602-NA Over-the-head 25 dB
19072315 MT53H7B4602-NA Neckband 25 dB
19072316 MT53H7P3E4602-NA Hard Hat 24 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19072314 Supplier 3M™ Supplier No. 7100204442

PELTOR™ LiteCom FRS Headset features a built-in analog two-way communication radio programmed to operate on the 22 FRS channels.

  • Protective communication solution for hands-free, clear communication in noisy environments
  • Analog two-way communication radio with 22 pre-programmed FRS channels
  • Noise-canceling microphone for clear speech transmission
  • Push-to-Talk (PTT) or hands-free Voice Operated (VOX) transmit options
  • Operates on AA batteries or rechargeable NiMH battery pack
  • Voice guided headset menu for easy operation
  • Comfortable for all day wearability

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Color Navy Blue
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Construction, Facility Maintenance, High Noise Environment, Machine Operations, Manufacturing
Model MT53H7A4602-NA
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type Protective Communication
Headband Color Black
Dielectric No
Style Over-the-head
Show More Show Less