Learn More
3M™ PELTOR™ Hygiene Kit for X Series Ear Muffs
3M™ PELTOR™ Hygiene Kit includes replaceable ear cushions and foam liners for X Series Ear Muffs
Supplier: 3M™ 7000104048
Description
- Hygiene kits consist of one pair of replacement cushions and foam inserts for 3M™ PELTOR™ X Series ear muffs (X1, X2, X3, X4, X5)
- 3M recommends replacing ear muff cushions and foam inserts every six months or whenever they become damaged.
- Kits are packaged 10 Each/Cs.
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
| Black | |
| Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding | |
| Hygiene Kit |
| PELTOR™ X4 Ear Muffs | |
| HYX4 | |
| Yes |