missing translation for 'onlineSavingsMsg'
Learn More

3M™ Peltor™ Earmuff Replacement Hygiene Kit for H10 Series

Catalog No. 18858P
Change view
Click to view available options
:
Hygiene Kit; 3M; For H10 Series; 20 kit/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18858P Hygiene Kit; 3M; For H10 Series; 20 kit/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18858P Supplier 3M™ Supplier No. HY10
Only null left

Extend the life of earmuffs with 1 pair of replacement cushions and 1 pair of dampers.

  • Compatible with the 3M™ Peltor™ Solutions
  • Works as adapter for Motorola Visar communication radio
  • Assists with clear, reliable communication in high-noise environments
  • Works in conjunction with 2-way radio for crisp 2-way communication
  • 3M Catalog# HY10

Specifications

Color Black
For Use With (Equipment) For H10 series
Model HY10
Product Type Hygiene Kit
Material ABS