Learn More
3M™ Peltor™ E·A·R· bud™ Noise Isolating Headphones Need - in E·A·R·
Description
- Made of foam plastic metal
- Provides NRR of 20dB with Ultrafit tips and NRR 29dB with the Skull Screw tips
- Volume output is limited to 82dB
- Available in black/gray color
- Assembly
- Demolition
- Electrical
- Facility maintenance
- Machine operations
- Remediation
- 3M Catalog# EARBUD2600N
Specifications
Specifications
| For Use With (Equipment) | Mobile phones and MP3 players that accept 3.5 mm plug |
| Model | EARBUD2600N |
| Noise Reduction Rating (NRR) | 26 dB (The NRR may overestimate the hearing protection provided during typical use. 3M recommends reducing the NRR by 50% for estimating the amount of noise reduction provided.) |
| Includes | One pair of the UltraFit™ Communication Eartips (NRR 20dB) and one pair of the Skull Screws™ Communication Eartips (NRR 29dB) |