missing translation for 'onlineSavingsMsg'
Learn More

3M™ TR-300 and TR-600 PAPR Battery Packs

Catalog No. p-57862
Change view
Click to view available options
For Use With (Equipment):
Breathe Easy™ PAPR Systems
Versaflo TR-600 PAPR Series
Color:
Blue
Blue, Gray
Battery Life:
8 to 18 hr.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Color Battery Life
296357897 Breathe Easy™ PAPR Systems - -
296358323 Breathe Easy™ PAPR Systems Blue -
19268020 Versaflo TR-600 PAPR Series Blue, Gray -
19268022 Versaflo TR-600 PAPR Series Blue, Gray 8 to 18 hr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296357897 Supplier 3M™ Supplier No. 7000131716
Only null left

Choose from a selection of lithium ion rechargeable batteries for the TR-300 or TR-600 PAPR systems.

  • Rechargeable or standard batteries
  • Choice of NiCd or Lithium
  • Long run time for filters and cartridges

Battery packs must be used in accordance with the user instructions, warnings and limitations accompanying each product

Compliance

NIOSH approved.

Specifications

Battery Type Lithium
Certifications/Compliance NIOSH Approved
Disposable Single-use
Product Type Battery Pack
For Use With (Equipment) Breathe Easy™ PAPR Systems