Learn More
3M™ OX™ Eyewear
Designed to fit over prescription glasses
$123.35 - $123.35
Specifications
| Certifications/Compliance | ANSI Z87.1-2015, CSA Z94.3 |
|---|---|
| Lens Tint | Clear |
| Frame Color | Black |
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Designed to fit over prescription glasses
- Seamless Duralite polycarbonate lens with built-in sideshields provides an uninterrupted sight range and optically correct peripheral vision
- Clear lens provides 99.9% UV protection
- Optional DX™ antifog hardcoat resists fogging, scratching, static and chemicals
- Choice of standard spatula temples or adjustable Secure-Grip™ temples with a pivoting axis for a secure, custom fit
- 3M Catalog# 12159-00000-20 , 3M Catalog# 12163-00000-20 , 3M Catalog# 12166-00000-20
Compliance
Meets ANSI Z87.1-2015 a nd CSA Z94.3 standards.
Specifications
| ANSI Z87.1-2015, CSA Z94.3 | |
| Clear | |
| Black |