Learn More
3M™ Nuvo™ Readers Eyewear
Catalog No. 296366552
Change view
Click to view available options
Lens Tint:
Clear
Magnification Power:
+1.50
Catalog No. Lens Tint Magnification Power
296366552 Clear +1.50
1 options
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 296366552 Supplier 3M™ Supplier No. 7000052797
Only null left
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Bifocal readers with three magnification diopters
Designed for workers who have difficulty reading small print or who are engaged in fine detail work.
  • Classic look of a dual lens but with tough, lightweight one-piece lens
  • Lens with anti-fog coating absorbs 99.9% UV
  • Gray, adjustable length temples with soft tips for comfortable fit
  • Soft nosepiece
  • Integral sideshields provide profile protection
  • Cushioned brow bar
  • 3M Catalog# 11434-00000-20, 11435-00000-20, 11436-00000-20, 11500-00000-20, 11501-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Bifocal Readers
For Use With (Application) Designed for workers who have difficulty reading small print or who are engaged in fine detail work
Magnification Power +1.50
Model 11434-00000-20
UV Protection UV400