missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Lens Cover
Description
- Used with the 3M wide-view lens Up to five covers can be attached, one on top of the other
- 3M Catalog# L-133-25
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
| Color | Clear |
| Disposable | Single-use |
| Product Type | Lens Cover |
| For Use With (Application) | Protects Lens From Scratching or Overspray of Paint and Chemicals |
| For Use With (Equipment) | 3 M™ Wide-View Lens of Powered and Supplied Air Respirator |
| Model | L-133-25 |
| Quantity | 25/Cs. |