Learn More
3M™ Hard Hat with Uvicator™ Sensor
Catalog No. p-4384237
Change view
Click to view available options
Color:
Black
Green
Ventilated:
Not Ventilated
Ventilated
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Ventilated
17623640 Green Not Ventilated
17623641 Green Ventilated
17623791 Black Not Ventilated
17623793 Black Ventilated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17623640 Supplier 3M™ Supplier No. 7000128175
Only null left
Protective hard hat with vents, comfortable 4-point ratchet suspension and standard brow pad
Available in a variety of colors, vented or unvented
  • Cap style hard hats constructed of HDPE
  • Four-point ratchet suspension
  • Uvicator lets wearer known when to replace hard hat due to UV exposure
  • Suspension sits lower on head to reduce pressure and increase security
  • Smooth, easy turning ratchet suspension
  • Dielectric: Class C, G, and E
  • 3M Catalog# H-701R-UV, H-701V-UV, H-702R-UV, H-702V-UV, H-703R-UV, H-703V-UV, H-704R-UV, H-704V-UV, H-709R-UV, H-711R-UV, H-711V-UV, H-712V-UV, H-713R-UV, H-713V-UV, H-712R-UV
Compliance
Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C.
Specifications
Color Green
Includes Uvicator
Model H-704R-UV
Product Type Hard Hat with Uvicator
Suspension Type 4-Point Ratchet
Ventilated Not Ventilated