Learn More
3M™ H-800 Series Full Brim Hard Hats
Catalog No. 17623706
Change view
Click to view available options
Color:
Gray
Hi-vis Orange
Hi-vis Yellow
Navy Blue
White
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17623706 White
17623712 Hi-vis Orange
17623713 Gray
17623714 Hi-vis Yellow
17623715 Navy Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17623706 Supplier 3M™ Supplier No. 7000144957
Only null left
Helps protect the head from small falling objects and fixed objects
  • Made of HDPE
  • Features 4-point ratchet suspension with three position height adjustment allows for customized fit
  • Perforated absorbent brow pad 
  • Optional 6-point suspension strap for more comfort
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
  • Available in several colors
  • 3M Catalog# H-801R, H-802R, H-803R, H-804R, H-805R, H-806R, H-807R, H-808R, H-809R, H-810R, H-811R, H-812R, H-813R
Specifications
Certifications/Compliance ANSI/ISEA Z89.1-2009 Type I, Class C, G and E
Color White
Includes 4-point Ratchet Suspension and Standard Brow Pad
Material HDPE
Model H-801R
Quantity 20/Cs.
Size (Metric) 168 to 197 mm
Suspension Type 4-point Ratchet
Weight (English) 416 oz.
Weight (Metric) 11.79 kg
Ventilated Not Ventilated
Head Style Full Brim
Show More Show Less