Learn More
3M™ H-700 Series Cap Style Hard Hats with 4-Point Ratchet Suspension
Catalog No. p-5211704
Change view
Click to view available options
Color:
Bright Yellow
Gray
Orange
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17623314 Gray
17623322 Bright Yellow
17623324 Orange
17623325 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17623314 Supplier 3M™ Supplier No. 7000030065
Only null left
Protect top of the head due to potential hazard of small falling objects or exposed fixed objects
  • Suspension adjusts easily to allow for customized height and fit
  • Replaceable brow pad for more comfort and extended shelf life
  • Short brim for upward visibility
  • Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
  • ANSI/ISEA compliant when worn in the forward and reverse donning positions
  • Available in various colors
  • 3M Catalog# H-701R, H-702R, H-703R, H-704R, H-705R, H-706R, H-707R, H-708R, H-709R, H-710R, H-711R, H-712R, H-713R
Specifications
Certifications/Compliance Meets ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
Color Gray
Includes Standard brow pad
Model H-708R
Product Type Cap-style Hard Hat
Quantity 20/Cs.
Suspension Type 4-Point Ratchet