missing translation for 'onlineSavingsMsg'
Learn More

3M™ Full Facepiece 6000DIN Series Respirator

Catalog No. p-322010
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19055521 Small
19055520 Medium
19055522 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19055521 Supplier 3M™ Supplier No. 7000126399
Only null left
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Requires

Filter media (sold separately).

Lightweight full-facepiece respirator with built-in DIN port adapter assembly features a well-balanced design and silicone faceseal for enhanced comfort, durability and ease of cleaning. Available in air-purifying, supplied air, and powered air purifying respirator modes.

  • Can be reconfigured for use with 2000 Series or 7093 filters, 6000 Series cartridges, and powered or supplied air systems against a variety of gases, vapors, and particulate hazards according to NIOSH approvals
  • Soft, lightweight, and well-balanced design
  • Adjustable head straps for long-wearing comfort
  • Large polycarbonate lens with scratch-resistant coating
  • No components made from natural rubber latex
  • 3M™ Cool Flow™ Valve helps make breathing easier
  • Available in air-purifying, supplied air, and powered air purifying respirator modes
  • Center adapter directs exhaled breath downward

Order Info

Facepieces and responder filter media sold separately. Consult your Fisher Safety Sales Representative for more information about appropriate uses and configurations.

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Compliance

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

ANSI Standard ANSI Z87.1
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Gray
Material Silicone, Thermoplastic Elastomer
Product Type Gas Mask Respirator
Size Small
Facepiece Material Silicone
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) 3M™ Filters 2000 Series, 3M™ Filter 7093, P100, 3M™ Cartridges 6000 Series
Includes Din port adapter assembly
Material (Lens) Polycarbonate
Model 6700DIN
Suspension Type 4-point
Show More Show Less