missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Filter Adapter 502
Description
The 3M™ Filter Adapter 502, Respiratory Protection System Component attaches 3M™ Particulate Filters 2000/2200 Series and 7093/7093C to the 3M Respirators 5000 Series and 3M Cartridges 6000 Series (all respirators, filters, cartridges, and adapters sold separately).
- Approved 3M system component
- Designed to attach 3M™ Particulate Filters 2000/2200 Series and 7093/7093C to 3M gas/vapor respirators and cartridges
- Component works with 3M Respirator 5000 Series and 3M Cartridges 6000 Series (sold separately)
- Particle filter adapters increases scope of potential applications
- Color: translucent white
- Refer to Filter/Cartridge User Instructions for more information
- 3M Catalog# 502
Specifications
Specifications
| Cartridge Type | Respirator Cartridge |
| Product Type | Adapter |
| For Use With (Equipment) | Self-contained Breathing Apparatus (SCBA) |