Learn More
3M™ E-A-R™ TaperFit™ 2 Ear Plugs
Catalog No. 19087852
Change view
Click to view available options
Model:
312-1224
Corded:
Corded
Size:
Large
Catalog No. Model Corded Size
19087852 312-1224 Corded Large
1 options
Catalog No. 19087852 Supplier 3M™ Supplier No. 7000002313
Only null left
Soft, roll-down foam ear plugs conform to the unique shape of each ear for a low-pressure, comfortable seal.
  • No-roll design keeps fingers, ear plugs, and ears clean
  • Slow recovery foam easily goes into ear canal and then sets and stays in place
  • Test compatible with 3M E-A-Rfit™ Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
ANSI S3.1974 Yes
Cord Color Blue
Cord Material Vinyl
CSA Z94.2 Class AL
Size Large
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 312-1224
Packaging Type Poly bag
Pairs per Package 200 Pairs/Pk.
Plug Material Foam
Plug Color Yellow
Noise Reduction Rating (NRR) 32 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Show More Show Less