Learn More
3M™ E-A-R™ Express™ Pod Plugs™ Ear Plugs
Catalog No. 18763
Change view
Click to view available options
Model:
311-1114
311-1115
321-2100
321-2200
Corded:
Corded
Uncorded
Product Type:
Assorted color grip ear plug
Blue grip ear plug
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Corded Product Type
18763 311-1115 Corded Assorted color grip ear plug
18999600 311-1114 Corded Blue grip ear plug
18586 321-2200 Uncorded Assorted color grip ear plug
18587 321-2100 Uncorded Blue grip ear plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18763 Supplier 3M™ Supplier No. 7000127179
Express™ Pod Plugs™ ear plugs allows the foam to compress so the ear plug slides gently into the ear and expands slowly. Flexible stem makes inserting easy.
  • Paddle-style grips provide quick, clean, and consistent placement of ear plugs
  • No-roll design keeps ear plugs and ears clean, even when fingers or gloves are dirty
  • Slow recovery foam easily goes into ear canal and then seats and stays in place
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
ANSI S3.1974 Yes
Cord Color Blue
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 311-1115
Packaging Type Pillow Pk.
Pairs per Package 400 Pairs/Pk.
Plug Material Foam, thermoplastic
Plug Color Yellow
Noise Reduction Rating (NRR) 25 dB
Corded Corded
Plug Shape Pod
Product Type Assorted color grip ear plug
Disposable Reusable
Metal Detectable No
Show More Show Less