missing translation for 'onlineSavingsMsg'
Learn More

3M™ P100 Comfort Plus 8293 Particulate Respirator

Catalog No. 19037352
Change view
Click to view available options
P100, Faceseal and Exhalation Valve
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19037352 P100, Faceseal and Exhalation Valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19037352 Supplier 3M™ Supplier No. 7000002055
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Lightweight P100 respirator is ideal for protection against oil based particles from sprays that do not also emit vapors.

  • NIOSH approved P100 respirator for solid particulates and liquid mists in concentrations not exceeding 10X PEL/OEL
  • Cool Flow™ exhalation valve reduces heat build-up inside the respirator
  • Compatible with a variety of protective eyewear and hearing protection
  • Adjustable noseclip helps provide a custom secure seal
  • Individually packaged
  • Advanced Electret Media is designed for ease of breathing

Specifications

Aerosol Type P - Oil-Proof
Assigned Protection Factor (APF) 10
Certifications (NIOSH) NIOSH 42 CFR 84, P100, approval number: TC-84A-2561
Certifications (OSHA) OSHA 1910.134
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type P100
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve Yes
Filter Efficiency 99.97%
For Use With (Application) Transportation, Construction, Mining, Oil & Gas, General Manufacturing
Latex Free Yes
Material Headstrap Polyisoprene, Cotton/Polyester
Model 8293
Nose Piece Material Aluminum
Nosepiece Type Nose clip
Packaging Type Individually packaged
Quantity 1 Ea.
Sterility Non-sterile
With Faceseal Yes
Strap Attachment Type Adjustable Buckle Straps
FDA Cleared Surgical Mask No
Show More Show Less