Learn More
3M™ Chemical Folded Sorbent Spill Kits
Use capillary action to draw in and trap liquids
Supplier: 3M™ 7000029637
Includes:
Compact chemroll kit includes one 25 ft. (7.6m) roll with a sorbing capacity of 5 gal. (19L) and a disposal bag. Large 31 gal. (117L) kit comes in a 20 gal. (76L) drum and includes 2 rolls and 30 pads. Each 50 ft. (15.2m) roll absorbs up to 10 1/2 gal. (40L). Measuring 17 x 19 in. (43.2 x 48.3cm) each, 30 pads absorb up to 111/4 gal. (42L).
Description
- These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
- 3M Catalog# CSKFL31 , 3M Catalog# CSKFL5
Specifications
| 20 gal. | |
| 75.7 L | |
| 20 gal. | |
| Chemical Sorbent Folded Spill Kit | |
| One 25 ft. roll with a sorbing capacity of 5 gal. and a disposal bag, large 31 gal. kit comes in a 20 gal. drum and includes 2 rolls and 30 pads, each 50 ft. roll absorbs up to 40L, measuring 17 x 19 in. each, 30 pads absorb up to 42L |
| 5 gal. | |
| 18.9 L | |
| 75.7 L | |
| For use on most aggressive, non aggressive liquids | |
| CSKFL5 |