Learn More
3M™ Centurion™ Impact Goggles
Catalog No. 19047712
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19047712 Anti-fog
1 options
Catalog No. 19047712 Supplier 3M™ Supplier No. 7100009575
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The durable 3M™ Centurion Impact Safety Goggle is reliable protective eyewear, featuring a wide-angle wraparound frame design with distinct 'side-to-side' peripheral vision and a large lens opening to accommodate prescription and safety eyewear with dust protection.
  • 3M Catalog# 40300-00000-10 , 3M Catalog# 40301-00000-10
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1 and CAN / CSA-Z94.3
Lens Tint Clear
Product Type Impact-resistant Goggles
For Use With (Equipment) Fits over most glasses
Material Polycarbonate
UV Protection UV400