Learn More
3M™ Breathing Tube for L-Series Loose-Fitting Headgear
Breathing tube to be used with L-Series headgear.
Supplier: 3M™ 7000002074
Includes: Assembly includes breathing tube, metal thumbscrew clamp, and squeeze clamp
Description
- Certain PAPR systems provide a higher level of respiratory protection than non-powered air filtering respirators
- Constant flow of air can provide cooling for the worker, resulting in greater comfort for the wearer
- Loose-fitting headgear options provide respiratory protection without the need for a tight faceseal or fit testing
- Battery-powered blower pulls the air through the filter resulting in no additional breathing resistance to the wearer
- 3M Catalog# L-122
Specifications
| Breathing Tube | |
| Assembly, grinding, machine operation | |
| L series headgear and helmet | |
| L-122 |