Learn More
3M™ Breathe-Easy™ Belt-Mounted PAPR System Breathing Tubes
Catalog No. 19805594
Change view
Click to view available options
For Use With (Equipment):
Breathe Easy™ powered air purifying respirator system, s series hood
Catalog No. For Use With (Equipment)
19805594 Breathe Easy™ powered air purifying respirator system, s series hood
1 options
All safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19805594 Supplier 3M™ Supplier No. 7100009877
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
All safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
Flexible, accordion-style hoses connect Turbo PAPR assembly to headgear.
3M Breathe Easy Belt-Mounted Turbo PAPRs
To assemble a complete system, select from the following pieces:
  • Headgear, facepiece, head cover or hood
  • Compatible breathing tube
  • Breathe Easy belt-mounted Turbo PAPR assembly
  • Appropriate cartridge/filter pack for your application
  • Battery pack and charger
  • 3M Catalog# 520-01-00R01 , 3M Catalog# 520-03-32R01 , 3M Catalog# BE-324
Order Info
Breathing tube clamps available. (See Cat. No. 19-813-724)
Specifications
Connection Type Belt Mounted
Material Vinyl
Product Type Breathing Tube
For Use With (Equipment) Breathe Easy™ powered air purifying respirator system, s series hood
Model BE-324