missing translation for 'onlineSavingsMsg'
Learn More

3M™ 6000 Series Resuable Half-Facepiece Respirators with Replaceable P100 Filters

Catalog No. p-56487
Change view
Click to view available options
For Use With (Application):
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding
Assembly, cleaning, grinding, machine operation, painting, welding
Format:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Format
19003456B Assembly, cleaning, grinding, machine operation, painting, welding Medium
19003456C Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, sanding, welding Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19003456B Supplier 3M™ Supplier No. 7000126163
Only null left

Lightweight, low-maintenance respirators come equipped with replaceable P100 filters (Style 2091)

  • Respirators can be disassembled, cleaned and reused; simply twist off and dispose of filters when they are used to capacity, and attach new filters
  • Effective for dusts, fumes, mists, radionuclides, radon daughters, and asbestos-containing dusts and mists
  • 3M Catalog# 6191 , 3M Catalog# 6291 , 3M Catalog# 6391

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Certifications

NIOSH 42 CFR 84 approved for use in areas where oil- and non-oil-based particles are present

Specifications

Certifications/Compliance NIOSH 42CFR84
Connection Type Bayonet
Material Thermoplastic Elastomer
Product Type Half Facepiece Respirator
Format Medium
For Use With (Application) Assembly, cleaning, grinding, machine operation, painting, welding
For Use With (Equipment) 6000 series reusable half facepiece respirator
Includes P100 filter
Model 6291