Learn More
Chemglass Inc 3000ml Reaction Vessel, Spherical, 100mm Schott O-Ring Flange, 185mm O.D. x 225mm Height
Catalog No. CG193202
Encompass
3000ml Reaction Vessel, Spherical, 100mm Schott O-Ring Flange, 185mm O.D. x 225mm Height. Spherical shaped reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Vessel will fit standard heating mantles CG-10000 through CG-12502. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.
Catalog No. CG193202 Supplier Chemglass Inc Supplier No. CG193202
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
3000ml Reaction Vessel, Spherical, 100mm Schott O-Ring Flange, 185mm O.D. x 225mm Height. Spherical shaped reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Vessel will fit standard heating mantles CG-10000 through CG-12502. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.