missing translation for 'onlineSavingsMsg'
Learn More

3H,5H-Octamethyltetrasiloxane, 96%

Catalog No. AAL1695614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1695614 25 g
AAL1695606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1695614 Supplier Thermo Scientific Chemicals Supplier No. L1695614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16066-09-4 | C8H24O3Si4 | 280.617 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16066-09-4
Molecular Formula C8H24O3Si4
Molecular Weight (g/mol) 280.617
MDL Number MFCD00053664
InChI Key OHSYWAVRSCQMHG-UHFFFAOYSA-N
Synonym 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, 3h,5h-octamethyltetrasiloxane, 1,3-bis trimethylsiloxy-1,3-dimethyldisiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl, bis trimethylsiloxy dimethyldisiloxane, methyl-methyl trimethylsilyloxy, 1,1,1,3,5,7,7,7-octamethyl-tetrasiloxane, tetrasiloxane, 1,1,1,3,5,7,7,7-octamethyl-,, 1,1,1,3,5,7,7,7-octamethyltetrasiloxane, technical gc
PubChem CID 6327318
IUPAC Name methyl-[methyl(trimethylsilyloxy)-$l^{3}-silanyl]oxy-trimethylsilyloxysilicon
SMILES C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 169°C
Quantity 25 g
Flash Point 78°C (172°F)
Formula Weight 282.64
Percent Purity 96%
Refractive Index 1.386
Sensitivity Moisture sensitive
Density 0.863
Chemical Name or Material 3H,5H-Octamethyltetrasiloxane
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 240-209-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only