missing translation for 'onlineSavingsMsg'
Learn More

3H-1,2-Benzodithiol-one 1,1-dioxide, 98%

Catalog No. p-7058536
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL20129MD 250 mg
AAL2012906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL20129MD Supplier Thermo Scientific Chemicals Supplier No. L20129MD
Only null left

CAS: 66304-01-6 | C7H4O3S2 | 200.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66304-01-6
Molecular Formula C7H4O3S2
Molecular Weight (g/mol) 200.226
MDL Number MFCD00132960
InChI Key JUDOLRSMWHVKGX-UHFFFAOYSA-N
PubChem CID 3009847
IUPAC Name 1,1-dioxo-1$l^{6},2-benzodithiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)SS2(=O)=O

Specifications

Melting Point 102°C to 107°C
Quantity 250 mg
UN Number UN3335
Beilstein 4180332
Synonym 3h-1,2-benzodithiol-3-one 1,1-dioxide, beaucage reagent, 3h-1,2-benzodithiol-3-one-1,1-dioxide, 3h-benzo c 1,2 dithiol-3-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one, 1,1-dioxide, 1,1-dioxo-1$l^ 6 ,2-benzodithiol-3-one, 3h-1,2-benzodithiol-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one1,1-dioxide, 1??,2-benzodithiole-1,1,3-trione, acmc-1b2sk
Formula Weight 200.24
Percent Purity 99%
Chemical Name or Material 3H-1,2-Benzodithiol-one 1,1-dioxide
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only