missing translation for 'onlineSavingsMsg'
Learn More

3H-1,2-Benzodithiol-3-one 1,1-Dioxide 98.0+%, TCI America™

Catalog No. B31251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31251G 1 g
B31255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31251G Supplier TCI America Supplier No. B31251G

Chemical Identifiers

CAS 66304-01-6
Molecular Formula C7H4O3S2
Molecular Weight (g/mol) 200.226
MDL Number MFCD00132960
InChI Key JUDOLRSMWHVKGX-UHFFFAOYSA-N
PubChem CID 3009847
IUPAC Name 1,1-dioxo-1$l^{6},2-benzodithiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)SS2(=O)=O

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 1 g
Synonym 3h-1,2-benzodithiol-3-one 1,1-dioxide, beaucage reagent, 3h-1,2-benzodithiol-3-one-1,1-dioxide, 3h-benzo c 1,2 dithiol-3-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one, 1,1-dioxide, 1,1-dioxo-1$l^ 6 ,2-benzodithiol-3-one, 3h-1,2-benzodithiol-one 1,1-dioxide, 3h-1,2-benzodithiol-3-one1,1-dioxide, 1??,2-benzodithiole-1,1,3-trione, acmc-1b2sk
Formula Weight 200.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3H-1,2-Benzodithiol-3-one 1,1-Dioxide