missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film

Single-use tank liners reduce cleaning steps common to re-usable tanks.
Supplier: Thermo Scientific™ B101459I
Description
The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Chemical Identifiers
| Polyethylene | |
| Individually packed in a header bag | |
| Room Temperature | |
| 0 Ports | |
| No Tubing |
| Store at room temperature. | |
| Irradiated | |
| HyPerforma | |
| ASI 26 |
Specifications
| 300 L | |
| Store at room temperature. | |
| Individually packed in a header bag | |
| Irradiated | |
| HyPerforma | |
| ASI 26 |
| Polyethylene | |
| 70 cm x 122 cm (24 in. x 48 in.) | |
| 300 L, 10 per case | |
| Room Temperature | |
| 0 Ports | |
| No Tubing |