missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film

Description
The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
| Capacity (Metric) | 100 L |
| Material | Polyethylene |
| Content And Storage | Store at room temperature. |
| Dimensions | 45.7 cm x 76.2 cm (18 in. x 30 in.) |
| Outer Packaging | Individually packed in a header bag |
| Quantity | 20 per case |
| Sterilization Method | Irradiated |
| Shipping Condition | Room Temperature |
| Product Line | HyPerforma |
| Port Detail | 0 Ports |
| Show More |