Learn More
Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film
GREENER_CHOICE
Single-use tank liners reduce cleaning steps common to re-usable tanks.
Supplier:  Thermo Scientific™ B100364I
 View more versions of this product
Catalog No. B100364I
Only null left
Description
Description
The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Chemical Identifiers
Polyethylene
Individually packed in a header bag
Room Temperature
0 Ports
No Tubing
Store at room temperature.
Irradiated
HyPerforma
ASI 26
Specifications
50 L
Store at room temperature.
Individually packed in a header bag
Irradiated
HyPerforma
ASI 26
Polyethylene
33 cm x 68.6 cm (13 in. x 27 in.)
40 per case
Room Temperature
0 Ports
No Tubing
Product Suggestions
Product Suggestions