missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film
GREENER_CHOICE

Catalog No. B100363I
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
300 L
560 L
Dimensions:
33 cm x 68.6 cm (13 in. x 27 in.)
45.7 cm x 76.2 cm (18 in. x 30 in.)
55.9 cm x 91.4 cm (22 in. x 36 in.)
70 cm x 122 cm (24 in. x 48 in.)
78.7 cm x 124.5 cm (31 in. x 49 in.)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Dimensions
B100363I 200 L 55.9 cm x 91.4 cm (22 in. x 36 in.)
B100364I 50 L 33 cm x 68.6 cm (13 in. x 27 in.)
B100373I 100 L 45.7 cm x 76.2 cm (18 in. x 30 in.)
B101459I 300 L 70 cm x 122 cm (24 in. x 48 in.)
B101460I 560 L 78.7 cm x 124.5 cm (31 in. x 49 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. B100363I Supplier Thermo Scientific™ Supplier No. B100363I

Single-use tank liners reduce cleaning steps common to re-usable tanks.

The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids

Specifications

Capacity (Metric) 200 L
Material Polyethylene
Content And Storage Store at room temperature.
Dimensions 55.9 cm x 91.4 cm (22 in. x 36 in.)
Outer Packaging Individually packed in a header bag
Quantity 20 per case
Sterilization Method Irradiated
Shipping Condition Room Temperature
Product Line HyPerforma
Port Detail 0 Ports
Film Description ASI 26
Tubing Style No Tubing
Show More Show Less