Learn More
Thermo Scientific™ 3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
Thermo Scientific 3D Tank Liners are composed of Aegis 5-14 film with dimensions optimized for Thermo Scientific drums and commonly-used, industry-standard cylindrical tanks. The tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
Supplier: Thermo Scientific™ SH3098803
Description
- Minimizes bioburden due to construction under sterile cGMP conditions
- Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
| 200 L | |
| 200 L | |
| C-Flex |